missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Granulated Media
- Formulation per liter: Casein Peptone [91079-40-2] 16g, Yeast Extract [8013-01-2] 10g, Sodium Chloride [7647-14-5] 5g
- Store at room temperature
Specifications
Specifications
| Form | Solid |
| Media Type | Dehydrated 2XYT |
| Product Type | Microbiology Media |
| Formulation | Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?