missing translation for 'onlineSavingsMsg'
Learn More

2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents

Product Code. 10123103
Change view
Click to view available options
Quantity:
10,000 g
100 g
2000 g
500 g
Packaging Type:
Poly Bottle
Poly Pail
Unit Size:
100g
10kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
10123103 10,000 g Poly Pail 10kg
10346793 100 g Poly Bottle 100g
10011223 2000 g Poly Bottle 2kg
10617633 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10123103 Supplier Fisher BioReagents Supplier No. BP246710
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use in molecular genetics

Formulation Usage: 31.0g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <4°C
Additive 2xyt Broth
Form Powder
Formulation 31g/L
For Use With (Application) Molecular Genetics
Packaging Type Poly Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.