missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents
Description
Formulation Usage: 31.0g/L
Specifications
Specifications
| Content And Storage | <4°C |
| Additive | 2xyt Broth |
| Form | Powder |
| Formulation | 31g/L |
| For Use With (Application) | Molecular Genetics |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?