missing translation for 'onlineSavingsMsg'
Learn More

3-(1,1,1-Trimethylhydrazin-1-ium-2-yl)propanoate Dihydrate, TRC

Product Code. 30133749
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133749 25 g 25g
30137048 5 g 5g
17944225 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133749 Supplier Toronto Research Chemicals Supplier No. TRCT89608025G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86426-17-7
Molecular Formula C6 H14 N2 O2 . 2 H2 O
Molecular Weight (g/mol) 182.22
Synonym 2-(2-Carboxyethyl)-1,1,1-trimethylhydrazinium inner salt hydrate (1:2), Hydrazinium, 2-(2-carboxyethyl)-1,1,1-trimethyl-, inner salt, dihydrate (9CI), 1,1,1-Trimethyl-2-(2-carboxyethyl)hydrazine zwitterion dihydrate, 3-(2,2,2-Trimethylhydrazinium)propionate dihydrate
IUPAC Name 3-[(trimethylazaniumyl)amino]propanoate;dihydrate
SMILES O.O.C[N+](C)(C)NCCC(=O)[O-]

Specifications

InChI Formula InChI=1S/C6H14N2O2.2H2O/c1-8(2,3)7-5-4-6(9)10;;/h7H,4-5H2,1-3H3;2*1H2
Quantity 25 g
Formula Weight 182.1267
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Meldonium dihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.