missing translation for 'onlineSavingsMsg'
Learn More

3-(1,1,2,3,3,3-Hexafluoropropyl)adamantane-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10623033
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10623033 1 g Amber Glass Bottle 1g
10520163 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10623033 Supplier Thermo Scientific Maybridge Supplier No. RJC02875DA
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 86301-98-6
Molecular Formula C14H16F6O2
Molecular Weight (g/mol) 330.27
MDL Number MFCD00193108
InChI Key GLJDZZQQPFAEPQ-GQPGPOQHSA-N
Synonym 1-1,1,2,3,3,3-hexafluoroprop-1-yl adamantane-3-carboxylic acid, 1r,3r,5r,7s-3-1,1,2,3,3,3-hexafluoropropyl adamantane-1-carboxylic acid, 1r,3s,5r,7s-3-1,1,2,3,3,3-hexafluoropropyl adamantane-1-carboxylic acid
PubChem CID 71299598
IUPAC Name (5S,7R)-3-(1,1,2,3,3,3-hexafluoropropyl)adamantane-1-carboxylic acid
SMILES C1C2CC3(CC1CC(C2)(C3)C(C(C(F)(F)F)F)(F)F)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 330.27
Percent Purity 97%
Chemical Name or Material 3-(1,1,2,3,3,3-Hexafluoropropyl)adamantane-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.