missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Benzothiazol-2-yl)aniline, Thermo Scientific™

Product Code. 10619705
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10619705

Brand: Thermo Scientific Maybridge MO08615DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 41230-21-1
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.297
InChI Key MKMRZZJJDIBAJF-UHFFFAOYSA-N
Synonym 3-benzothiazol-2-yl-phenylamine, 3-1,3-benzothiazol-2-yl aniline, 3-benzothiazol-2-ylphenylamine, benzenamine,3-2-benzothiazolyl, cambridge id 5250055, 3-2-benzothiazolyl aniline, 3-benzothiazol-2-yl aniline, 3-benzo d thiazol-2-yl aniline, 3-benzothiazol-2-yl benzenamine, 3-1,3-benzothiazol-2-yl phenyl amine
PubChem CID 720999
IUPAC Name 3-(1,3-benzothiazol-2-yl)aniline
SMILES C1=CC=C2C(=C1)N=C(S2)C3=CC(=CC=C3)N

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.3
Percent Purity 97%
Chemical Name or Material 3-(1, 3-benzothiazol-2-yl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.