missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10506134
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10506134 250 mg Amber Glass Bottle 250mg
10225704 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10506134 Supplier Thermo Scientific Maybridge Supplier No. CC58801CB
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-44-2
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.21
MDL Number MFCD09702391
InChI Key ZODQJPSGAHBVHU-UHFFFAOYSA-N
Synonym 3-1,3-dioxolan-2-yl thiophene-2-carboxylic acid, 3-1,3-dioxolan-2-yl-2-thiophenecarboxylic acid, 2-2-carboxythien-3-yl-1,3-dioxolane, 3-1,3-dioxolan-2-yl thiophene-2-carboxylicacid, 2-thiophenecarboxylicacid, 3-1,3-dioxolan-2-yl
PubChem CID 24229593
IUPAC Name 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
SMILES OC(=O)C1=C(C=CS1)C1OCCO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.21
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Dioxolan-2-yl)-2-thiophenecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.