missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dioxolan-2-yl)thiophene-2-sulfonyl chloride, ≥97%, Thermo Scientific™

Product Code. 10061574
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10061574 1 g Amber Glass Bottle 1g
10051574 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061574 Supplier Thermo Scientific Maybridge Supplier No. MO07810DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 103011-38-7
Molecular Formula C7H7ClO4S2
Molecular Weight (g/mol) 254.70
MDL Number MFCD08271969
InChI Key RRTOURREASJQSR-UHFFFAOYSA-N
Synonym 3-1,3-dioxolan-2-yl thiophene-2-sulfonyl chloride, chloro 3-1,3-dioxolan-2-yl 2-thienyl sulfone, 3-1,3-dioxolan-2-yl-thiophene-2-sulfonyl chloride
PubChem CID 10538915
IUPAC Name 3-(1,3-dioxolan-2-yl)thiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=C(C=CS1)C1OCCO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 254.71
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Dioxolan-2-yl)thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.