missing translation for 'onlineSavingsMsg'
Learn More

3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%

Product Code. 15424097
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424097 5 g 5g
15414097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424097 Supplier Thermo Scientific Alfa Aesar Supplier No. H31465.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 87151-66-4 | C9H14O | 138.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87151-66-4
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD12407081
InChI Key DJJNAQALDCPMOU-UHFFFAOYSA-N
Synonym 3-1,4-cyclohexadien-1-yl-1-propanol, 3-cyclohexa-1,4-dien-1-yl propan-1-ol, 3-1,4-cyclohexadienyl propan-1-ol, 3-3-hydroxypropyl-1,4-cyclohexadiene
PubChem CID 20402847
IUPAC Name 3-cyclohexa-1,4-dien-1-ylpropan-1-ol
SMILES C1C=CCC(=C1)CCCO

Specifications

Quantity 5 g
Sensitivity Air sensitive
Formula Weight 138.21
Percent Purity 97%
Chemical Name or Material 3-(1,4-Cyclohexadien-1-yl)-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.