missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Azepanyl)-2,2-dimethylpropylamine, ≥95%, Thermo Scientific™

Product Code. 10551573
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10551573 250 mg 250mg
10503983 1 g 1g
10662875 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10551573 Supplier Thermo Scientific Maybridge Supplier No. MO01190CB
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-85-0
Molecular Formula C11H24N2
Molecular Weight (g/mol) 184.327
MDL Number MFCD06200874
InChI Key SUDFIJJDSKVMSO-UHFFFAOYSA-N
Synonym 3-azepan-1-yl-2,2-dimethylpropan-1-amine, 3-1-azepanyl-2,2-dimethylpropylamine, 3-azepan-1-yl-2,2-dimethyl-propylamine, 1h-azepine-1-propanamine,hexahydro-b,b-dimethyl, 3-azaperhydroepinyl-2,2-dimethylpropylamine
PubChem CID 2794705
IUPAC Name 3-(azepan-1-yl)-2,2-dimethylpropan-1-amine
SMILES CC(C)(CN)CN1CCCCCC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 184.32
Percent Purity 95%
Chemical Name or Material 3-(1-Azepanyl)-2, 2-dimethylpropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.