missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10245854
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10245854

Brand: Thermo Scientific Maybridge CC60516DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 179055-95-9
Molecular Formula C11H9N3
Molecular Weight (g/mol) 183.21
MDL Number MFCD09702401
InChI Key NEFOVTAGRMOIIV-UHFFFAOYSA-N
PubChem CID 10511671
IUPAC Name 3-(2-methylpyrazol-3-yl)benzonitrile
SMILES CN1N=CC=C1C1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-1-methyl-1h-pyrazol-5-yl benzonitrile, 3-2-methylpyrazol-3-yl benzonitrile, benzonitrile,3-1-methyl-1h-pyrazol-5-yl, benzonitrile, 3-1-methyl-1h-pyrazol-5-yl, 3-1-methylpyrazol-5-yl benzenecarbonitrile
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.