missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10255854
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10255854 5 g Amber Glass Bottle 5g
10245854 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255854 Supplier Thermo Scientific Maybridge Supplier No. CC60516DE
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 179055-95-9
Molecular Formula C11H9N3
Molecular Weight (g/mol) 183.21
MDL Number MFCD09702401
InChI Key NEFOVTAGRMOIIV-UHFFFAOYSA-N
PubChem CID 10511671
IUPAC Name 3-(2-methylpyrazol-3-yl)benzonitrile
SMILES CN1N=CC=C1C1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 3-1-methyl-1h-pyrazol-5-yl benzonitrile, 3-2-methylpyrazol-3-yl benzonitrile, benzonitrile,3-1-methyl-1h-pyrazol-5-yl, benzonitrile, 3-1-methyl-1h-pyrazol-5-yl, 3-1-methylpyrazol-5-yl benzenecarbonitrile
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.