missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Pyridinio)-1-propanesulfonate, 99%

Product Code. 15557039 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15557039 250 g 250g
15547039 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15557039 Supplier Thermo Scientific Acros Supplier No. 459672500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15471-17-7 | C8H11NO3S | 201.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15471-17-7
Molecular Formula C8H11NO3S
Molecular Weight (g/mol) 201.24
MDL Number MFCD00064468
InChI Key REEBJQTUIJTGAL-UHFFFAOYSA-N
Synonym 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium
PubChem CID 84929
IUPAC Name 3-pyridin-1-ium-1-ylpropane-1-sulfonate
SMILES [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1

Specifications

Melting Point 275.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 250 g
Solubility Information (10% in water) Clear solution
Formula Weight 201.25
Percent Purity 99%
Chemical Name or Material 3-(1-Pyridinio)-1-propanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.