missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Pyrrolyl)benzoic acid, 97%

Product Code. 15465917
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465917 10 g 10g
15455917 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465917 Supplier Thermo Scientific Alfa Aesar Supplier No. H33124.09
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61471-45-2 | C11H8NO2 | 186.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61471-45-2
Molecular Formula C11H8NO2
Molecular Weight (g/mol) 186.19
MDL Number MFCD02656610
InChI Key PODFNQCZFHLJPH-UHFFFAOYSA-M
Synonym 3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
PubChem CID 736537
IUPAC Name 3-pyrrol-1-ylbenzoic acid
SMILES [O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1

Specifications

Melting Point 175°C to 178°C
Quantity 10 g
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1-Pyrrolyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.