missing translation for 'onlineSavingsMsg'
Learn More

(3β,16α)-3,16-Dihydroxypregn-5-en-20-one, TRC

Product Code. 30386236
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386236 Supplier Toronto Research Chemicals Supplier No. TRCD455615100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3beta,16alpha)-3,16-Dihydroxypregn-5-en-20-one
CAS 520-88-7
Molecular Formula C21 H32 O3
Quantity 100 mg
Synonym 16α-Hydroxypregnenolone, 3β,16α-Dihydroxypreg-5-en-20-one, 3β,16α-Dihydroxypregnen-5-en-20-one, Pregn-5-en-3β,16α-diol-20-one
InChI Formula InChI=1S/C21H32O3/c1-12(22)19-18(24)11-17-15-5-4-13-10-14(23)6-8-20(13,2)16(15)7-9-21(17,19)3/h4,14-19,23-24H,5-11H2,1-3H3/t14-,15-,16+,17+,18-,19+,20+,21-/m1/s1
SMILES CC(=O)[C@H]1[C@H](O)C[C@H]2[C@@H]3CC=C4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@@]12C
IUPAC Name 1-[(3R,8S,9S,10R,13R,14S,16R,17R)-3,16-dihydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
Molecular Weight (g/mol) 332.477
Formula Weight 332.235
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.