missing translation for 'onlineSavingsMsg'
Learn More

3-(1h-1,2,4-Triazol-1-yl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10184033
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184033 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10184033 Supplier Thermo Scientific Maybridge Supplier No. CC38404DA
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 868755-54-8
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H7N3O
MDL Number MFCD08271896
Quantity 1 g
Synonym 3-1h-1,2,4-triazol-1-yl benzaldehyde, 3-1,2,4-triazol-1-yl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-yl, pubchem22932, 3-1,2,4-triazolyl benzaldehyde, 3-1,2,4 triazol-1-yl-benzaldehyde
InChI Key YGKBHMJLDGOYPI-UHFFFAOYSA-N
SMILES O=CC1=CC(=CC=C1)N1C=NC=N1
IUPAC Name 3-(1,2,4-triazol-1-yl)benzaldehyde
Molecular Weight (g/mol) 173.18
PubChem CID 18525741
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.