missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10178182
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10178182 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10178182 Supplier Thermo Scientific Maybridge Supplier No. CC45401DA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 857284-23-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9N3O2
MDL Number MFCD07186456
Quantity 1 g
Synonym 3-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 3-1,2,4 triazol-1-ylmethyl-benzoic acid, 3-1,2,4-triazol-1-ylmethyl benzoic acid, benzoic acid,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazolylmethyl benzoic acid, 3-1h-1,2,4-triazol-1-yl methyl benzoic acid, benzoic acid, 3-1h-1,2,4-triazol-1-ylmethyl
InChI Key XIJPFLBFQZEFKY-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(CN2C=NC=N2)=CC=C1
IUPAC Name 3-(1,2,4-triazol-1-ylmethyl)benzoic acid
Molecular Weight (g/mol) 203.20
PubChem CID 3159713
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-triazol-1-ylmethyl)benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.