missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-imidazol-1-yl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10892221
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10892221 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10892221 Supplier Thermo Scientific Maybridge Supplier No. MAY00311DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 127404-22-2
Assay Percent Range 97%
Molecular Formula C10H8N2O
Quantity 1 g
Synonym 3-1h-imidazol-1-yl benzaldehyde, 3-1-imidazolyl benzaldehyde, benzaldehyde, 3-1h-imidazol-1-yl, 3-imidazol-1-yl benzaldehyde, pubchem22683, 3-imidazolylbenzaldehyde
InChI Key CGTQCZGDVDTRDI-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)N2C=CN=C2)C=O
IUPAC Name 3-imidazol-1-ylbenzaldehyde
Molecular Weight (g/mol) 172.187
PubChem CID 15650408
Formula Weight 172.19
Chemical Name or Material 3-(1H-imidazol-1-yl)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.