missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-yl)benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10611465
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10611465 10 g Amber Glass Bottle 10g
10601465 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10611465 Supplier Thermo Scientific Maybridge Supplier No. CC32916EA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25699-82-5
Molecular Formula C10H7N3
Molecular Weight (g/mol) 169.19
MDL Number MFCD07772807
InChI Key KLZKYUMVLIQOFX-UHFFFAOYSA-N
PubChem CID 7164581
SMILES N#CC1=CC(=CC=C1)N1C=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3-1h-pyrazol-1-yl benzonitrile, 3-pyrazol-1-yl benzonitrile, 3-pyrazolylbenzenecarbonitrile, 3-1h-pyrazole-1-yl benzonitrile, benzonitrile,3-1h-pyrazol-1-yl, benzonitrile, 3-1h-pyrazol-1-yl
IUPAC Name 3-(1H-pyrazol-1-yl)benzonitrile
Formula Weight 169.19
Percent Purity 97%
Chemical Name or Material 3-(1H-pyrazol-1-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.