missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)benzaldehyde, 95+%, Thermo Scientific™

Product Code. 10702103
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10702103 250 mg 250mg
10029121 1 g 1g
10652305 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702103 Supplier Thermo Scientific Maybridge Supplier No. CC21904CB
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 129747-77-9
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.20
MDL Number MFCD03086140
InChI Key PALTUANHIBXQMX-UHFFFAOYSA-N
PubChem CID 2776528
IUPAC Name 3-pyrrol-1-ylbenzaldehyde
SMILES O=CC1=CC(=CC=C1)N1C=CC=C1

Specifications

Assay Percent Range 95%
Quantity 250 mg
Synonym 3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde
Formula Weight 171.2
Percent Purity 95%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.