missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%

Product Code. 11436396
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436396 500 g 500g
11416396 25 g 25g
11426396 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436396 Supplier Thermo Scientific Alfa Aesar Supplier No. B23159.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3069-29-2 | C8H22N2O2Si | 206.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3069-29-2
Molecular Formula C8H22N2O2Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00054776
InChI Key MQWFLKHKWJMCEN-UHFFFAOYSA-N
Synonym n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
PubChem CID 76482
IUPAC Name N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine
SMILES CO[Si](C)(CCCNCCN)OC

Specifica

Boiling Point 265°C
Quantity 500 g
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.449
Sensitivity Air and moisture sensitive
Density 0.968
UN Number UN2735
Beilstein 2074259
Formula Weight 206.36
Percent Purity 97%
Chemical Name or Material 3-(2-Aminoethylamino)propylmethyldimethoxysilane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.