missing translation for 'onlineSavingsMsg'
Learn More

3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide, TRC

Product Code. 30308896
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308896 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308896 Supplier Toronto Research Chemicals Supplier No. TRCA6115255MG
 Restricted Item
Estimated Shipment: 10-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 3-[((2-[(Diaminomethylene)-amino]thiazol-4-yl)methyl)sulfanyl]-propanamide
CAS 76824-16-3
Purity Grade Notes HPLC
Molecular Formula C8 H13 N5 O S2
Quantity 5 mg
Synonym Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]- (9CI), 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfanyl]-propanamide, Famotidine Imp. D (EP)
InChI Formula InChI=1S/C8H13N5OS2/c9-6(14)1-2-15-3-5-4-16-8(12-5)13-7(10)11/h4H,1-3H2,(H2,9,14)(H4,10,11,12,13)
SMILES NC(=Nc1nc(CSCCC(=O)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanamide
Molecular Weight (g/mol) 259.35
Formula Weight 259.0562
Percent Purity >95
Mostrar más Mostrar menos
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.