missing translation for 'onlineSavingsMsg'
Learn More

3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile, Thermo Scientific™

Product Code. 10717115
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10717115 25 g 25g
10565943 1 g 1g
10575943 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10717115 Supplier Thermo Scientific Maybridge Supplier No. CD05258EB
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-65-9
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
InChI Key NVZISNDFOXXQSQ-UHFFFAOYSA-N
Synonym 3-2-bromo-5-methoxyphenyl-3-oxopropanenitrile, maybridge1_002847, 3-2-bromo-5-methoxy-phenyl-3-oxo-propanenitrile
PubChem CID 2804251
IUPAC Name 3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile
SMILES COC1=CC(=C(C=C1)Br)C(=O)CC#N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 254.08
Percent Purity 97%
Chemical Name or Material 3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.