missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloroethyl)-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, TRC

Product Code. 30137450
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137450 50 mg 50mg
30009992 25 mg 25mg
30006058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137450 Supplier Toronto Research Chemicals Supplier No. TRCC36627050MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130049-82-0
Molecular Formula C11 H15 Cl N2 O2
Molecular Weight (g/mol) 242.7
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-chloroethyl)-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-, 3-(2-Chloroethyl)-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-Chloroethyl)-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-Chloroethyl)-9-hydroxy-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 9-Hydroxy-3-(2-chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
IUPAC Name 3-(2-chloroethyl)-9-hydroxy-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(CCCl)C(=O)N2CCCC(O)C2=N1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H15ClN2O2/c1-7-8(4-5-12)11(16)14-6-2-3-9(15)10(14)13-7/h9,15H,2-6H2,1H3
Formula Weight 242.0822
Chemical Name or Material 3-(2-Chloroethyl)-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrimidin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.