missing translation for 'onlineSavingsMsg'
Learn More

3-(2-chlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10364582
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10364582 10 g Amber Glass Bottle 10g
10727305 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10364582 Supplier Thermo Scientific Maybridge Supplier No. SB01885EA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 23598-72-3
Molecular Formula C11H8ClNO3
Molecular Weight (g/mol) 237.64
MDL Number MFCD00020813
InChI Key UVEPOHNXGXVOJE-UHFFFAOYSA-N
Synonym 3-2-chlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2'-chlorophenyl-4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl, 3-2-chloro-phenyl-5-methyl-isoxazole-4-carboxylic acid, 3-2-chlorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-o-chlorophenyl-5-methyl, 4-carboxy-3-2-chlorophenyl-5-methylisoxazole, 3 2-chlorophenyl-5-methyl-isoxazolyl-4-carboxylic acid, 5-methyl-3-2-chlorophenyl-4-isoxazolecarboxylic acid
PubChem CID 90203
IUPAC Name 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl

Specifications

CAS Min % 97.0
Quantity 10 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 237.64
Percent Purity 97%
Chemical Name or Material 3-(2-chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.