missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)-1-methyl-1H-pyrazole-5-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10787475
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10787475 1 g Amber Glass Bottle 1g
10507854 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10787475 Supplier Thermo Scientific Maybridge Supplier No. CC47001DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 859851-00-6
Molecular Formula C9H8N2O3
Molecular Weight (g/mol) 192.17
MDL Number MFCD08271950
InChI Key MOPCEJWYTICWJM-UHFFFAOYSA-N
Synonym 3-2-furyl-1-methyl-1h-pyrazole-5-carboxylic acid, 5-furan-2-yl-2-methylpyrazole-3-carboxylic acid, 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl, 3-fur-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1-methylpyrazole-5-carboxylic acid
PubChem CID 7537643
SMILES CN1N=C(C=C1C(O)=O)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 3-(furan-2-yl)-1-methyl-1H-pyrazole-5-carboxylic acid
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)-1-methyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.