missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10412694
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10412694 1 g Amber Glass Bottle 1g
10402694 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412694 Supplier Thermo Scientific Maybridge Supplier No. CC46101DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 35461-99-5
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD02690979
InChI Key RQVVFGRDMHDHNI-UHFFFAOYSA-N
Synonym 3-furan-2-yl benzoic acid, 3-2-furyl benzoic acid, 3-fur-2-ylbenzoic acid, 3-furan-2-yl-benzoic acid, m-2-furyl benzoic acid, benzoic acid, 3-2-furanyl
PubChem CID 5037933
IUPAC Name 3-(furan-2-yl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.