missing translation for 'onlineSavingsMsg'
Learn More

3-[(2-Hydroxyethyl)thio]-6-nitro-2,3-dihydro-1H-1λ 6-benzo[b]thiophene-1,1-diol, 97%, Thermo Scientific™

Product Code. 10334672
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10334672 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10334672 Supplier Thermo Scientific Maybridge Supplier No. KM08337DA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175203-70-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11NO5S2
MDL Number MFCD00203984
Quantity 1 g
Synonym 3-2-hydroxyethyl thio-6-nitro-2,3-dihydro-1h-1lambda∼6∼-benzo b thiophene-1,1-dione, 2-6-nitro-1,1-dioxido-2,3-dihydro-1-benzothien-3-yl thio ethanol, 3-2-hydroxyethyl sulfanyl-6-nitro-2,3-dihydro-1??-benzothiophene-1,1-dione, maybridge1_005013, 2-6-nitro-1,1-dioxo-2,3-dihydro-1-benzothiophen-3-yl sulfanyl ethanol, 2-6-nitro-1,1-dioxo-2,3-dihydro-1-benzothiophen-3-yl thio ethanol, 3-2-hydroxyethyl thio-6-nitro-2,3-dihydrobenzo b thiophene 1,1-dioxide, ethanol,2-2,3-dihydro-6-nitro-1,1-dioxidobenzo b thien-3-yl thio, 2-6-nitro-1,1-bis oxidanylidene-2,3-dihydro-1-benzothiophen-3-yl sulfanyl ethanol, 3-2-hydroxyethyl sulfanyl-6-nitro-2,3-dihydro-1$l^ 6-benzothiophene-1,1-dione
InChI Key FLAITYNRCKZPST-UHFFFAOYSA-N
SMILES C1C(C2=C(S1(=O)=O)C=C(C=C2)[N+](=O)[O-])SCCO
IUPAC Name 2-[(6-nitro-1,1-dioxo-2,3-dihydro-1-benzothiophen-3-yl)sulfanyl]ethanol
Molecular Weight (g/mol) 289.32
PubChem CID 2779237
Formula Weight 289.33
Percent Purity 97%
Chemical Name or Material 3-[(2-Hydroxyethyl)thio]-6-nitro-2, 3-dihydro-1H-1lambda∼6∼-benzo[b]thiophene-1, 1-dio
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.