missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10304472
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10304472

Brand: Thermo Scientific Maybridge CC19401CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 28077-41-0
Molecular Formula C11H9NO2S
Molecular Weight (g/mol) 219.258
MDL Number MFCD02682065
InChI Key YDPHSMPSNLAMJE-UHFFFAOYSA-N
Synonym 3-2-methyl-1,3-thiazol-4-yl benzoic acid, 3-2-methylthiazol-4-yl benzoic acid, benzoic acid,3-2-methyl-4-thiazolyl, 3-2-methyl thiazol-4-yl benzoic acid
PubChem CID 2776495
IUPAC Name 3-(2-methyl-1,3-thiazol-4-yl)benzoic acid
SMILES CC1=NC(=CS1)C2=CC(=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-1, 3-thiazol-4-yl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.