missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Oxo-2,3-dihydro-1,3-benzoxazol-3-yl)propanoic acid, 97%, Thermo Scientific™

Product Code. 10305112
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10305112 10 g Amber Glass Bottle 10g
10273232 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10305112 Supplier Thermo Scientific Maybridge Supplier No. SPB02696EA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13610-59-8
Molecular Formula C10H9NO4
Molecular Weight (g/mol) 207.185
MDL Number MFCD00052166
InChI Key BMLRDVCKJXMASM-UHFFFAOYSA-N
Synonym 3-2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl propanoic acid, 3-2-oxo-1,3-benzoxazol-3 2h-yl propanoic acid, 3-2-oxobenzo d oxazol-3 2h-yl propanoic acid, 3-2-oxo-benzooxazol-3-yl propionic acid, 3-2-oxo-1,3-benzoxazol-3-yl propanoic acid, 3 2h-benzoxazolepropanoic acid, 2-oxo, 3-2-oxo-benzooxazol-3-yl-propionic acid, 3 2h-benzoxazolepropanoicacid, 2-oxo, 3-2-oxo-3-hydrobenzoxazol-3-yl propanoic acid, maybridge1_008314
PubChem CID 720147
IUPAC Name 3-(2-oxo-1,3-benzoxazol-3-yl)propanoic acid
SMILES C1=CC=C2C(=C1)N(C(=O)O2)CCC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 207.18
Percent Purity 97%
Chemical Name or Material 3-(2-Oxo-2, 3-dihydro-1, 3-benzoxazol-3-yl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.