missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)-L-alanine, 95%

Product Code. 15479948
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15479948 1 g 1g
15499948 250 mg 250mg
15489948 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15479948

Brand: Thermo Scientific Alfa Aesar H63316.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22951-96-8 | C7H9NO2S | 171.214 g/mol

3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22951-96-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00064335
InChI Key WTOFYLAWDLQMBZ-LURJTMIESA-N
Synonym 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715
PubChem CID 146719
IUPAC Name (2S)-2-amino-3-thiophen-2-ylpropanoic acid
SMILES C1=CSC(=C1)CC(C(=O)O)N

Specifications

Melting Point 255°C to 263°C (decomposition)
Color White
Beilstein 82872
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 171.21
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(2-Thienyl)-L-alanine
compliance-icons
Product Identifier
  • 3-(2-Thienyl)-L-alanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-(2-Thienyl)-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.