missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester, 97%

Product Code. 15434486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434486 5 g 5g
15424486 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434486 Supplier Thermo Scientific Alfa Aesar Supplier No. H58731.06
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 915402-03-8 | C17H25BO2Si | 300.28 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915402-03-8
Molecular Formula C17H25BO2Si
Molecular Weight (g/mol) 300.28
MDL Number MFCD08706285
InChI Key LDKJXCOLGXLWKP-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-3-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-trimethylsilane, trimethyl 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 3-trimethylsilyl ethynyl phenylboronic acid pinacol ester, trimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, amtb201, 3-2-trimethylsilyl ethynyl benzeneboronic acid pinacol ester
PubChem CID 17999151
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C#C[Si](C)(C)C

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
IUPAC Name trimethyl({2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl})silane
Formula Weight 300.28
Percent Purity 97%
Chemical Name or Material 3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.