missing translation for 'onlineSavingsMsg'
Learn More

3,3',3' '-(2,4,6-boroxintriyl)tris-Pyridine Hydrochloride (>70%), TRC

Product Code. 30421919
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421919 1 g 1g
30414323 100 mg 100mg
30411566 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421919 Supplier Toronto Research Chemicals Supplier No. TRCP9970251G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3',3''-(2,4,6-boroxintriyl)tris-Pyridine Hydrochloride
Molecular Formula C15H12B3N3O3 •xHCl
Quantity 1 g
Synonym 3,3',3''-(2,4,6-Boroxintriyl)tris[pyridine] Hydrochloride, 2,4,6-Tris(3-pyridinyl)cyclotriboroxane Hydrochloride, Tris(3-pyridyl)boroxin Hydrochloride
InChI Formula InChI=1S/C15H12B3N3O3.ClH/c1-4-13(10-19-7-1)16-22-17(14-5-2-8-20-11-14)24-18(23-16)15-6-3-9-21-12-15;/h1-12H;1H
SMILES B1(OB(OB(O1)C2=CC=CN=C2)C3=CC=CN=C3)C4=CN=CC=C4.Cl
IUPAC Name 2,4,6-tri(pyridin-3-yl)-1,3,5,2,4,6-trioxatriborinane hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.