missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-1-propanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11435600
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11435600

Brand: Thermo Scientific Alfa Aesar H25856.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845866-80-0
Molecular Formula C3H4ClF3O2S
Molecular Weight (g/mol) 196.568
MDL Number MFCD06248898
InChI Key OXKHPRFGCFJUSK-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-1-propanesulfonyl chloride, 3,3,3-tris fluoranyl propane-1-sulfonyl chloride, chloro 3,3,3-trifluoropropyl sulfone, 3,3,3-trifluoropropylsulfonyl chloride, 3,3,3-trifluoropropanesulfonyl chloride, 3,3,3-trifluoropropylsulphonyl chloride, 3,3,3-trifluoropropane sulfonyl chloride, 3,3,3-trifluoropropane-1-sulphonyl chloride, 1-propanesulfonylchloride, 3,3,3-trifluoro, 3,3,3-trifluoro-1-propane sulfonyl chloride
PubChem CID 2761177
IUPAC Name 3,3,3-trifluoropropane-1-sulfonyl chloride
SMILES C(CS(=O)(=O)Cl)C(F)(F)F

Specifications

Boiling Point 77°C to 78°C (16 mmHg)
Linear Formula CF3(CH2)2SO2Cl
Refractive Index 1.3997
Quantity 500 mg
UN Number UN3265
Sensitivity Air and moisture sensitive
Formula Weight 196.57
Percent Purity 95%
Chemical Name or Material 3,3,3-Trifluoro-1-propanesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.