missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoropropionic acid, 98%

Product Code. 10548833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10548833 1 mL Glass Bottle 1mL
10015063 5 mL Glass Bottle 5mL
10127143 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10548833 Supplier Thermo Scientific Acros Supplier No. 381070010
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2516-99-6 | C3H3F3O2 | 128.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-99-6
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD00153292
InChI Key KSNKQSPJFRQSEI-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid
PubChem CID 2777972
IUPAC Name 3,3,3-trifluoropropanoic acid
SMILES C(C(=O)O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 9.7°C
Color Colorless
Density 1.4500g/mL
Boiling Point 146°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula F3CCH2COOH
Refractive Index 1.3320 to 1.334
Quantity 1 mL
Specific Gravity 1.45
Formula Weight 128.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 3, 3-Trifluoropropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,3,3-Trifluoropropionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.