missing translation for 'onlineSavingsMsg'
Learn More

(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, Thermo Scientific Chemicals

Product Code. 10656650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656650 5 g Glass bottle 5g
10795832 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656650 Supplier Thermo Scientific Acros Supplier No. 432210050
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1481-41-0
Molecular Formula C5H10ClF3Si
Molecular Weight (g/mol) 190.67
InChI Key KBAZUXSLKGQRJF-UHFFFAOYSA-N
Synonym trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
PubChem CID 73866
IUPAC Name chloro-dimethyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](C)(CCC(F)(F)F)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.1100g/mL
Boiling Point 118°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.3770 to 1.3790 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.11
Formula Weight 190.67
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (3, 3, 3-Trifluoropropyl)chlorodimethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (3,3,3-Trifluoropropyl)chlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.