missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Benzophenonetetracraboxylic Dianhydride, TRC

Product Code. 30331783
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331783 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331783 Supplier Toronto Research Chemicals Supplier No. TRCB1303905G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3',4,4'-Benzophenonetetracraboxylic Dianhydride
CAS 2421-28-5
Molecular Formula C17H6O7
Quantity 5 g
Synonym 5,5'-Carbonylbis[1,3-isobenzofurandione, 3,3',4,4'-Benzophenonetetracarboxylic Acid Dianhydride, 3,3',4,4'-Benzophenonetetracarboxylic Anhydride, 3,3',4,4'-Benzophenonetetracarboxylic Dianhydride, 3,3',4,4'-Tetracarboxybenzophenone Dianhydride, 3,4,3',4'-Benzophenonetetracarboxylic Acid Anhydride, 3,4,3',4'-Benzophenonetetracarboxylic Dianhydride, 4,4'-Carbonylbis(phthalic Anhydride), 4,4'-Carbonyldiphthalic Anhydride, 4,4'-Diphthalic Anhydride Ketone, BTDA, BTDA-PF, BTDA-UP, Benzophenone-3,3',4,4'-tetracarboxylic Acid Anhydride, Benzophenonetetracarboxylic Acid Dianhydride, Benzophenonetetracarboxylic Dianhydride, NSC 78480
InChI Formula InChI=1S/C17H6O7/c18-13(7-1-3-9-11(5-7)16(21)23-14(9)19)8-2-4-10-12(6-8)17(22)24-15(10)20/h1-6H
SMILES O=C(c1ccc2C(=O)OC(=O)c2c1)c3ccc4C(=O)OC(=O)c4c3
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 322.22
Formula Weight 322.0114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.