missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine, TRC

Product Code. 30453025
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453025 25 g -
30449190 10 g 10g
30455719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453025 Supplier Toronto Research Chemicals Supplier No. TRCT88954025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16 H20 N2
Molecular Weight (g/mol) 240.34
Synonym [1,1'-Biphenyl]-4,4'-diamine, 3,3',5,5'-tetramethyl- (9CI, ACI), 3,3',5,5'-Tetramethyl[1,1'-biphenyl]-4,4'-diamine (ACI), Benzidine, 3,3',5,5'-tetramethyl- (6CI), 3,3',5,5'-Tetramehylbenzidine, 3,3',5,5'-Tetramethyl-4,4'-diaminobiphenyl, 3,3',5,5'-Tetramethylbenzidine, 3,3',5,5'-Tetramethylbiphenyl-4,4'-diamine, 3,5,3',5'-Tetramethylbenzidine, 4,4'-Diamino-3,3',5,5'-tetramethyl-1,1'-biphenyl, BM blue, ColorBurst Blue, Enhanced K-Blue, K-Blue, K-Blue Max, Sure Blue TMB, TMB, TMB Blotting Plus, TMB substrate
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES Cc1cc(cc(C)c1N)c2cc(C)c(N)c(C)c2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3
Formula Weight 240.1626
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.