missing translation for 'onlineSavingsMsg'
Learn More

3,3’,5-Triiodo-L-thyronine Methyl Ester, TRC

Product Code. 30451196
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451196 25 mg 25mg
30457139 50 mg -
30455251 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451196 Supplier Toronto Research Chemicals Supplier No. TRCT79537325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54914-53-3
Molecular Formula C16 H14 I3 N O4
Molecular Weight (g/mol) 665
Synonym O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-L-Tyrosine methyl ester, 3,5,3'-Triiodo-L-thyronine methyl ester, L-Triiodothyronine methyl ester
IUPAC Name methyl (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
SMILES COC(=O)[C@@H](N)Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H14I3NO4/c1-23-16(22)13(20)6-8-4-11(18)15(12(19)5-8)24-9-2-3-14(21)10(17)7-9/h2-5,7,13,21H,6,20H2,1H3/t13-/m0/s1
Formula Weight 664.8057
Chemical Name or Material Liothyronine Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.