missing translation for 'onlineSavingsMsg'
Learn More

3,3’,5-Triiodo-L-thyronine Methyl Ester Trifluoroacetamide, TRC

Product Code. 30456880
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456880 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456880 Supplier Toronto Research Chemicals Supplier No. TRCT79540350MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3',5-Triiodo-L-thyronine Methyl Ester Trifluoroacetamide
CAS 3006-04-0
Molecular Formula C18 H13 F3 I3 N O5
Quantity 50 mg
Synonym (S)-Methyl 3-(4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl)-2-(2,2,2-trifluoroacetamido)propanoate, L-3-[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-N-(trifluoroacetyl)-alanine Methyl Ester
InChI Formula InChI=1S/C18H13F3I3NO5/c1-29-16(27)13(25-17(28)18(19,20)21)6-8-4-11(23)15(12(24)5-8)30-9-2-3-14(26)10(22)7-9/h2-5,7,13,26H,6H2,1H3,(H,25,28)/t13-/m0/s1
SMILES COC(=O)[C@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)NC(=O)C(F)(F)F
IUPAC Name methyl (2S)-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-2-[(2,2,2-trifluoroacetyl)amino]propanoate
Molecular Weight (g/mol) 761.008
Formula Weight 760.788
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.