missing translation for 'onlineSavingsMsg'
Learn More

3,3',5-Triiodo-L-thyronine sodium salt

Product Code. p-7042672
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 15415579

Brand: Thermo Scientific Alfa Aesar J63312.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55-06-1 | C15H11I3NNaO4 | 672.959 g/mol

L-3,3',5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-3,3′,5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.

Solubility
Partly miscible in water. Soluble in dimethyl sulfoxide, methanol, sodium hydroxide and ethanol. Practically insoluble in most other organic solvents.

Notes
Store at -20°C. Incompatible with oxidizing agents and heat. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-06-1
Molecular Formula C15H11I3NNaO4
Molecular Weight (g/mol) 672.959
MDL Number MFCD00002594
InChI Key SBXXSUDPJJJJLC-YDALLXLXSA-M
Synonym liothyronine sodium, cytomel, triostat, tertroxin, 3,3',5-triiodo-l-thyronine sodium salt, basoprocin, ibiothyron, cynomel, cytomel sodium, triiodothyronine sodium
PubChem CID 23666110
ChEBI CHEBI:6484
IUPAC Name sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
SMILES C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)[O-])N)I)I)O.[Na+]

Specifications

Melting Point 205°C
Color Brown to White
Quantity 500 mg
Beilstein 8179867
Sensitivity Light sensitive
Merck Index 14,5510
Solubility Information Partly miscible in water. Soluble in dimethyl sulfoxide,methanol,sodium hydroxide and ethanol. Practically insoluble in most other organic solvents.
Formula Weight 672.96
Physical Form Powder
Chemical Name or Material 3,3',5-Triiodo-L-thyronine sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.