missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-n-propylthiacarbocyanine iodide, 96%, Thermo Scientific Chemicals

Product Code. 15425127
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425127 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15425127 Supplier Thermo Scientific Alfa Aesar Supplier No. H32400.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53336-12-2
Molecular Formula C23H25IN2S2
Molecular Weight (g/mol) 520.49
MDL Number MFCD00013340
InChI Key JGTCEHAVVINOPG-UHFFFAOYSA-M
Synonym 3,3'-dipropylthiacarbocyanine iodide, 3-propyl-2-3-3-propyl-2 3h benzothiazolylidene-1-propenyl benzothiazolium iodide, 3-propyl-2-1e,3z-3-3-propylbenzo d thiazol-2 3h-ylidene prop-1-en-1-yl benzo d thiazol-3-ium iodide
PubChem CID 57369736
SMILES [I-].CCCN1C(SC2=CC=CC=C12)=CC=CC1=[N+](CCC)C2=CC=CC=C2S1

Specifications

Quantity 250 mg
IUPAC Name 3-propyl-2-[3-(3-propyl-1,3-benzothiazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzothiazole;iodide
Formula Weight 520.5
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-propylthiacarbocyanine iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.