missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392

Product Code. 15487195
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487195 100 g 100g
15477195 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487195 Supplier Thermo Scientific Alfa Aesar Supplier No. 046403.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1071466-61-9 | C25H28O4 | 392.495 g/mol

It is used in epoxy systems or in combination with acrylates, methacrylates, and maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071466-61-9
Molecular Formula C25H28O4
Molecular Weight (g/mol) 392.495
MDL Number MFCD22200539
InChI Key GJTZQXNRNNDGRG-UHFFFAOYSA-N
Synonym 2,2'-diallyl bisphenol a diacetate ether, 4,4'-isopropylidene bis 2-allylphenol acetate, phenyl ester epoxy curative hybrid of bisphenol a, 3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392, 4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate
PubChem CID 59215472
IUPAC Name [4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetate
SMILES CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C

Specifications

Color Amber
Physical Form Amber liquid
Quantity 100 g
Formula Weight 392.5
Chemical Name or Material 3,3'-Diallylbisphenol A diacetate, Epoxy curative, EC-392

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.