missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11378906
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378906 1 g 1g
11388906 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378906 Supplier Thermo Scientific Alfa Aesar Supplier No. L19532.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165904-27-8
Molecular Formula C13H27BO4
Molecular Weight (g/mol) 258.165
MDL Number MFCD03788723
InChI Key VEDSPUPKZZMMLL-UHFFFAOYSA-N
Synonym 3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
PubChem CID 12115795
IUPAC Name 2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC

Specifications

Refractive Index 1.428
Quantity 1 g
Formula Weight 258.17
Percent Purity 97%
Chemical Name or Material 3,3-Diethoxy-1-propylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.