missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylthiadicarbocyanine perchlorate, 96%, Thermo Scientific™

Product Code. 15448085
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448085 250 mg 250mg
15495137 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448085 Supplier Thermo Scientific Alfa Aesar Supplier No. H32421.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22268-65-1
Molecular Formula C23H23ClN2O4S2
Molecular Weight (g/mol) 491.017
MDL Number MFCD06409265
InChI Key HSVLFBSXAKAPBG-UHFFFAOYSA-M
Synonym benzothiazolium, 3-ethyl-2-5-3-ethyl-2 3h-benzothiazolylidene-1,3-pentadien-1-yl-, perchlorate 1:1, 3,3'-diethyl-2,2'-thiadicarbocyanine perchlorate, 3-ethyl-2-1e,3e,5e-5-3-ethylbenzo d thiazol-2 3h-ylidene penta-1,3-dien-1-yl benzo d thiazol-3-ium perchlorate, 3-ethyl-2-1e,3e-5-2e-3-ethyl-1,3-benzothiazol-2-ylidene penta-1,3-dien-1-yl-1,3-benzothiazol-3-ium perchlorate
PubChem CID 57357756
IUPAC Name 3-ethyl-2-[5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;perchlorate
SMILES CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.[O-]Cl(=O)(=O)=O

Specifications

Quantity 250 mg
Formula Weight 491.12
Percent Purity 96%
Chemical Name or Material 3,3'-Diethylthiadicarbocyanine perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.