missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diindolylmethane, 96%, Thermo Scientific™

Product Code. 15502363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15502363 25 g 25g
15592353 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15502363 Supplier Thermo Scientific Acros Supplier No. 459220250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1968-05-4
Molecular Formula C17H14N2
Molecular Weight (g/mol) 246.31

Specifications

Melting Point 164.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 25 g
Synonym heptane, 3-bromo, 3-heptyl bromide, acmc-1bscs
InChI Key MLHXKYLLJRLHGH-UHFFFAOYNA-N
SMILES CCCCC(CC)Br
IUPAC Name 3-bromoheptane
PubChem CID 16091
Formula Weight 246.31
Percent Purity 96%
Chemical Name or Material 3,3'-Diindolylmethane
Show More Show Less
compliance-icons
Product Identifier
  • 3,3'-Diindolylmethane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.