missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diindolylmethane, TRC

Product Code. 30381696
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381696 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381696 Supplier Toronto Research Chemicals Supplier No. TRCD455190100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1968-05-4
Molecular Formula C17 H14 N2
Quantity 100 mg
Synonym 1H-Indole, 3,3'-methylenebis-, Indole, 3,3'-methylenedi- (6CI,7CI,8CI), 3,3'-Methylenebis[1H-indole], 3,3'-Bisindolylmethane, 3,3'-Diindolylmethane, Arundine, Bis(1H-indol-3-yl)methane, Bis(3-indolyl)methane, Cervicon-DIM, DIM (AhR agonist), DIM (diindolylmethane), Diindolylmethane, HB 236, Infemin
InChI Formula InChI=1S/C17H14N2/c1-3-7-16-14(5-1)12(10-18-16)9-13-11-19-17-8-4-2-6-15(13)17/h1-8,10-11,18-19H,9H2
SMILES C(c1c[nH]c2ccccc12)c3c[nH]c4ccccc34
IUPAC Name 3-(1H-indol-3-ylmethyl)-1H-indole
Molecular Weight (g/mol) 246.31
Formula Weight 246.1157
Chemical Name or Material 3,3'-Di-Indoylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.