missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethoxyestra-5(10),9(11)-dien-17-one, TRC

Product Code. 30008609
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008609 100 mg 100mg
30007687 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008609 Supplier Toronto Research Chemicals Supplier No. TRCD476605100MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10109-76-9
Molecular Formula C20 H28 O3
Molecular Weight (g/mol) 316.43
Synonym Estra-5(10),9(11)-diene-3,17-dione 3-(Dimethyl Acetal), (8S,13S,14S)-3,3-dimethoxy-13-methyl-3,4,6,7,8,12,13,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17(2H)-one;
IUPAC Name (8S,13S,14S)-3,3-dimethoxy-13-methyl-1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-one
SMILES COC1(CCC2=C(CC[C@H]3[C@@H]4CCC(=O)[C@@]4(C)CC=C23)C1)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H28O3/c1-19-10-8-15-14-9-11-20(22-2,23-3)12-13(14)4-5-16(15)17(19)6-7-18(19)21/h8,16-17H,4-7,9-12H2,1-3H3/t16-,17+,19+/m1/s1
Formula Weight 316.2038
Chemical Name or Material 3,3-Dimethoxyestra-5(10),9(11)-dien-17-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.