missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1,2-epoxybutane, 95%

Product Code. 15478859
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478859 5 g 5g
11357066 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478859 Supplier Thermo Scientific Alfa Aesar Supplier No. L06951.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2245-30-9 | C6H12O | 100.161 g/mol

3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-30-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00051590
InChI Key HEAYDCIZOFDHRM-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1,2-epoxybutane, tert-butyloxirane, tert-butylethylene oxide, 3,3-dimethylbutene oxide, oxirane, 1,1-dimethylethyl, 3,3-dimethyl-1-butene oxide, 1,2-epoxy-3,3-dimethylbutane, tert.-butyloxirane, tert-butyl oxirane, butane, 1,2-epoxy-3,3-dimethyl
PubChem CID 92174
IUPAC Name 2-tert-butyloxirane
SMILES CC(C)(C)C1CO1

Specifications

Density 0.822
Boiling Point 95°C to 96°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.401
Quantity 5 g
UN Number UN1993
Beilstein 102478
Formula Weight 100.16
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1,2-epoxybutane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethyl-1,2-epoxybutane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butyloxirane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.