missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-(3-nitrophenyl-2,4,5,6-d4)triaz-1-ene, TRC

Product Code. 30179899
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179899 10 mg 10mg
30179759 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179899 Supplier Toronto Research Chemicals Supplier No. TRCD15346210MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2519481-67-3
Molecular Formula C8 D4 H6 N4 O2
Molecular Weight (g/mol) 198.215
Synonym 1-(m-Nitrophenyl)-3,3-dimethyltriazine-d4, 3,3-Dimethyl-1-(3-nitrophenyl)triazene-d4, NSC 82309-d4
IUPAC Name N-methyl-N-[(E)-(2,3,4,6-tetradeuterio-5-nitrophenyl)diazenyl]methanamine
SMILES [2H]c1c([2H])c(\N=N\N(C)C)c([2H])c(c1[2H])N(=O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H10N4O2/c1-11(2)10-9-7-4-3-5-8(6-7)12(13)14/h3-6H,1-2H3/b10-9+/i3D,4D,5D,6D
Formula Weight 198.105
Chemical Name or Material 3,3-Dimethyl-1-(3-Nitrophenyl-2,4,5,6-D4)Triaz-1-Ene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.